Masterflex® Peristaltic Pumps and Fluid Handling Solutions
Carousel
Masterflex® peristaltic pump systems provide superior accuracy and repeatability to ensure optimal performance when used with the proper tubing. These pumps offer the latest technology, including MasterSense™ integrated sensor monitoring to ensure media quality, MasterflexLive® cloud-enabled connectivity, intuitive digital touch screen capability and the low-maintenance and cost-effectiveness of peristaltic pumps. Choose from a wide range of flows with the Masterflex Ismatec Series, Masterflex L/S Series, Masterflex I/P Series and Masterflex B/T Series and many tubing formulations available, our extensive portfolio has what you need to scale from lab to production
Choose from a wide range of flows with our varied peristaltic pump series:
- Masterflex Ismatec® Series 0.0001 to 365 mL/min
- Masterflex L/S® Series 0.00005 to 3400 mL/min
- Masterflex I/P® Series 0.001 to 26 LPM
- Masterflex B/T® Series 0.3 to 42 LPM
With over 10,000 pump options and many tubing formulations, you’ll be able to find just what you need from the Masterflex® brand.
Masterflex® peristaltic pump systems, engineered for a long life, provide superior accuracy and repeatability, ensuring optimal performance from the research phase to scaled-up production.
Our pumps boast the latest in biometrics technology, including MasterflexLive® cloud-enabled connectivity, easy control and tubing changes — a low-maintenance and cost-effective solution to your peristaltic pump needs.
Choose from a wide range of flows with our varied peristaltic pump series:
- Masterflex Ismatec® series 0.0001 to 365 ml/min
- Masterflex L/S® series 0.00005 to 3400 ml/min
- Masterflex I/P® series 0.001 to 26 LPM
- Masterflex B/T® series 0.3 to 42 LPM
With over 10,000 pump options and many tubing formulations, you’ll be able to find just what you need from the Masterflex® brand.
Top products for biopharma and beyond
Peristaltic pumps
MasterSense™ smart pump systems
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Find your pumpFluid Handling Solutions Features & Benefits
Our fluid handling solutions are designed to support your entire enterprise. Our system supports traceability and IQ/OQ/PQ.
Our pumps have long been trusted for:
- Bioprocessing
- Chemical feed
- Food and beverage
- Cosmetic dispensing
- Inks/coatings
- Pulp and paper
- Semiconductors
- Water and wastewater
Furthermore, our pumps feature unparalleled benefits, including:
- Tough proprietary design that doesn’t break down in brutal environments
- Top quality for accuracy, precision and repeatability
- High volumetric system in which fluid touches only tubing material
Fluid Handling Solutions — From Research to Production
Dedicated to the success of your entire fluid path – from research to production
Masterflex® is dedicated to your success, from laboratory research to full-scale production. Our rugged pumps and tubing last, meaning fewer interruptions during the scientific process, as well as reliable accuracy and full compliance with standards, measures and regulations.
Featured Products
Custom Services and Tools
Custom engineered solutions
Partnering on custom engineered pumps – making ideas reality for over 50 years.
Learn more
Custom 'cut to length' tubing
We have the flexibility to meet your needs and save you time and money. Choose your tubing type, length and quantity.
Learn more
MasterflexLive®
With MasterflexLive®, you get real time monitoring of all pump parameters.
Learn more
Chemical compatibility
Search for your chemical to see the effect it has on hundreds of materials
Learn more
Masterflex Selection Tool
Find the recommended bundled peristaltic pump system for you in less than six questions!
Learn more
Connect with Masterflex®, Part of Avantor
Connect with Masterflex® (part of Avantor) to stay up to date on advanced and innovative fluid path solutions with:
- Case studies
- Webinars
- Guides and how-tos
We are always expanding our product lines to better serve you. See all Masterflex products.