Contact Plates & Dipslides
Contact plates available in blister, double or triple wrapped packaging complete with dipslides, all according to regulations. Available with TLHTh (tween, lecithin, histidine, thiosulphate) neutraliser accompanied by a certificate of irradiation.
Settle plates are irradiated for active or passive air monitoring. They come in blister or classical packaging with dipslides, all according to regulations.
Product Selection
Contact Plates
A simple and efficient way to detect microbiological contaminations on surfaces.
Dipslides
Testing with dip slides is a safe, reliable , economical and fast indicator of the presence of bacteria, yeasts and moulds on surfaces and in liquids...
Mini Incubator
Handy mini incubator for hygiene testing dipslides and contact plates.