Titanium ≥99.4% (metals basis), foil, annealed, Thickness: 0.74 mm (0.029 in)

Supplier: Thermo Fisher Scientific
Warning

00029.GX 00029.ZP
00029.GXEA 194 EUR
00029.GX 00029.ZP
Titanium ≥99.4% (metals basis), foil, annealed, Thickness: 0.74 mm (0.029 in)
Titanium

Purity: 99.4+% CAS No.: 7440-32-6 Physical Form: ≈200g/200x300mm Appearance: Dark gray Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR