Yttrium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Fisher Scientific


Packaged under argon.

00615.09 00615.18
00615.09EA 160 EUR
00615.09 00615.18
Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
Yttrium

Purity: 99.9% CAS No.: 7440-65-5 Physical Form: Packaged under argon Odour: Odourless MDL No.: MFCD00011468

Formula: Y
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR