Titanium ≥99% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Fisher Scientific
Warning

00971.FIEA 54.4 EUR
00971.FI 00971.GT
Titanium ≥99% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Titanium

Purity: 99% CAS No.: 7440-32-6 Physical Form: ≈1.4g/50x50mm Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR