Dextran sulphate sodium salt, M.W. ~ 500,000 ≥98%, Ultra Pure Grade

Supplier: VWR Chemicals
Warning

0198-250GEA 914 EUR
0198-250G 0198-50G 0198-2.5KG
Dextran sulphate sodium salt, M.W. ~ 500,000 ≥98%, Ultra Pure Grade
Dextran sulphate sodium salt

Used for the detection of DNA or RNA in nucleic acid hybridisation procedures. It effectively “excludes” probe from solution, accelerating the rate of annealing.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Refrigerator
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Specification Test Results

DNase none detected
Heavy Metals = 0.005 %
RNase none detected
Iron = 0.001 %
Loss on Drying = 10 %
pH (1%, Water) @25°C 6.4 - 8
Protease none
Purity 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR