Sodium dodecyl sulphate (SDS) ≥99%, Biotechnology Grade for biotechnology

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

0227-1KGEA 765 EUR
0227-1KG 0227-100G 0227-250G 0227-500G 0227-12KG 0227-5KG
Sodium dodecyl sulphate (SDS) ≥99%, Biotechnology Grade for biotechnology
Sodium dodecyl sulphate (SDS)

For applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@260nm (3%, Water) <0.1
Abs.@280nm (3%, Water) ≤0.1
Chloride ≤0.1%
Copper ≤0.0005%
DNase none detected
Heavy Metals (as Pb) ≤0.0005%
Iron ≤0.0001%
Phosphate ≤0.0005%
Protease none detected
Purity ≥99%
RNase none detected
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR