Sodium nitrite ≥97%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Diazotizing salts

0297.0100EA 34.89 EUR
0297.0100 0297.1000
Sodium nitrite ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium nitrite
Formula: NaNO₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Order Now

Specification Test Results

Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs.
Assay min. 97.0 %
Calcium (Ca) max. 0.01 %
Chloride (Cl) max. 0.005 %
Potassium (K) max. 0.005 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.01 %
Sulfate (SO4) max. 0.01 %
Trace Impurities (in ppm)
Arsenic (As) max. 3
Iron (Fe) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR