Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor

Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate

0304.1000EA 59.27 EUR
0304.1000 0304.9050
Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
di-Sodium hydrogen phosphate dodecahydrate
Formula: Na₂HPO₄·12H₂O
Melting Pt: 35 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733

Order Now

Specification Test Results

Assay 98.0 - 101.0 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 8.7 - 9.3
Sulfate (SO4) max. 0.02 %
Trace Impurities (in ppm)
Nitrogen Compounds (as N) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR