Ammonium carbonate, Ultrapure
Supplier: VWR Chemicals
Warning
Synonyms: Salt of hartshorn
0305-1KGEA 131 EUR
0305-1KG
Ammonium carbonate, Ultrapure
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: 58 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508
Order Now
Specification Test Results
Assay (NH3) >30%
Chloride <0.0005%
Heavy Metals (as Pb) <0.0005%
Insolubles <0.005%
Iron <0.0005%
Non-volatile Matter <0.01%
Sulphur Compounds <0.002%