Sodium deoxycholate 99,0, high_purity

Supplier: VWR Chemicals
Warning

Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate

0613-100GEA 218 EUR
0613-100G 0613-50G 0613-500G
Sodium deoxycholate 99,0, high_purity
Sodium deoxycholate

Anionic - negatively-charged (carboxylates, sulphates or sulphonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.

Formula: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7

Order Now

Specification Test Results

Arsenic <= 0.001 %
Heavy Metals <= 0.005 %
Loss on Drying <= 5.0 %
Purity (Dry Basis) 99.0%
Sodium Cholate <= 2.0 %
Solubility (10%, water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR