7,8-Dimethylalloxazine, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Lumichrome, 7,8-Dimethylbenzo[g]pteridine-2,4(1H,3H)-dione

103217-1G 103217-5G
103217-1GEA 149 EUR
103217-1G 103217-5G
7,8-Dimethylalloxazine, Sigma-Aldrich®
7,8-Dimethylalloxazine

Lumichrome (7,8-Dimethylalloxazine) is a natural metabolite of riboflavin. It is a flourescent photoproduct of riboflavin degradation. It is known to be an effective photosensitiser and a fluorescent dye. Lumichrome is suitable as a fluorescence probe, metal ion sensor, and may be used for the inactivation of biological contaminants. Its applications are also seen in memory and semiconductor devices.

Formula: C₁₂H₁₀N₄O₂
Melting Pt: >300 °C (lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00005021
CAS Number: 1086-80-2
EINECS: 214-120-8

Order Now

Specification Test Results

Appearance (Colour) Yellow
Appearance (Form); Powder, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Wavelength 214 - 224 nm
Extinction Coefficient ≥23000
TLC Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR