Tin ≥99.8% (metals basis), shot 8-20 mesh

Supplier: Thermo Fisher Scientific

10377.22 10377.36 10377.A3
10377.22EA 74 EUR
10377.22 10377.36 10377.A3
Tin ≥99.8% (metals basis), shot 8-20 mesh
Tin

Purity: 99.8% CAS No.: 7440-31-5 Odour: Odourless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR