Titanium ≥99.99% (metals basis), cubes 9.5x9.5x9.5 mm (0.37x0.37x0.37 in)

Supplier: Thermo Fisher Scientific
Warning

10380.KG 10380.KJ
10380.KGEA 78.9 EUR
10380.KG 10380.KJ
Titanium ≥99.99% (metals basis), cubes 9.5x9.5x9.5 mm (0.37x0.37x0.37 in)
Titanium

Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈3.9g/cube Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR