trans-Platin ≥64,5%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)

10472.01 10472.03 10472.06
10472.01EA 84.3 EUR
10472.01 10472.03 10472.06
trans-Platin ≥64,5%
trans-Platin

CAS No.: 14913-33-8 Molecular Formula: Cl2H6N2Pt Formula Weight: 300.06 Physical Form: Crystalline Appearance: Yellow Odour: Odourless MDL No.: MFCD00011623

Formula: H₆Cl₂N₂Pt
Melting Pt: 340 °C (dec.)
Density: 3.746 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 14913-33-8
EINECS: 238-980-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR