Iron(II) oxalate dihydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate

10718.22 10718.14
10718.22EA 136 EUR
10718.22 10718.14
Iron(II) oxalate dihydrate ≥99.999% (metals basis), Puratronic®
Iron (II) oxalate dihydrate

Purity: 99.999% CAS No.: 6047-25-2 Molecular Formula: FeC2O4.2H2O Formula Weight: 179.90 (143.87anhy) Physical Form: Powder Density: 2.28 Melting Point: 190° dec. MDL No.: MFCD00150040

Formula: Fe(C₂O₄)·2H₂O
Melting Pt: 190 °C (dec.)
Density: 2.28
Storage Temperature: Ambient
MDL Number: MFCD00150040
CAS Number: 6047-25-2
EINECS: 208-217-4
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR