Tin hard ≥99.9% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Fisher Scientific

10789.VAEA 214 EUR
10789.VA 10789.VE
Tin hard ≥99.9% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)
Tin

Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈4.2g/15x15cm, Temper: as rolled Odour: Odourless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR