Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium iodide hydrate

10871.06 10871.14 10871.22
10871.06EA 51.5 EUR
10871.06 10871.14 10871.22
Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®
Sodium iodide dihydrate

Purity: 99.999% Molecular Formula: NaI.xH2O Formula Weight: 149.89(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic

Formula: INa·2H₂O
Melting Pt: 64.3 °C
Density: 2.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149211
CAS Number: 13517-06-1
EINECS: 231-679-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR