Calcium oxide ≥99.95% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

10923.18 10923.30 10923.A1
10923.18EA 79.1 EUR
10923.18 10923.30 10923.A1
Calcium oxide ≥99.95% (metals basis)
Calcium oxide

Purity: 99.95% CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: 10 Micron APS Powder Density: 3.37 Refractive Index: 1.83 Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 2572° Boiling Point: 2850° Merck Reference: 14,1686 MDL No.: MFCD00010911

Formula: CaO
Boiling Pt: 2850 °C
Melting Pt: 2572 °C
Density: 3.37
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 14,1686

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR