Cyclobutanone 98+%

Supplier: Thermo Fisher Scientific
Danger

110930010 110930050 110930250
110930010.EA 56.6 EUR
110930010. 110930050. 110930250.
Cyclobutanone 98+%
Cyclobutanone
Formula: C₄H₆O
Boiling Pt: 99 °C (760 mmHg)
Melting Pt: –50.9 °C
Density: 0.93 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001332
CAS Number: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4230 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR