VWR® Vienna, Safety Spectacles
Supplier: VWR Collection
111-0931EA 10 EUR
111-0931
VWR® Vienna, Safety Spectacles
Safety Glasses
Lightweight safety glasses with clear PC lens.
  • Anti-scratch/anti-fog coating
  • Pantoscopic adjustment for angle modification
  • Moulded brow guard
  • Adjustable temple length
Weight: 26 g
EN 166, EN 170; 1 FT
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens).