Banded earplugs, 1310
Supplier: 3M
1310 1311 ES01300 ES01301 ES01301UK ES01300UK
111-1031EA 116 EUR
111-1031 111-1032 111-0445 111-0446 111-1074 111-1688
Banded earplugs, 1310
Earplugs Banded Earplugs
These earplugs are ideal for long periods of use with changing noise levels.
  • Flexible band design helps maintain a constant yet comfortable low pressure irrespective of head size
  • Can be worn behind the head or under the chin
  • Round plugs do not enter the ear canal for better hygiene
  • Replacement plugs are economical and easy to fit
SNR: 26 dB
EN 352-2
Certifications: EN 352-2