Bilsom™ 303 Disposable Earplugs
Supplier: HONEYWELL SAFETY
1006186 1006187 1006202 1005073 1005074 1007192 1007193 1006204
111-1291EA 50.8 EUR
111-1291 111-1292 111-1287 111-3560 111-3564 111-2157 111-2158 DALL1006204
Bilsom™ 303 Disposable Earplugs
Earplugs Disposable Earplugs
Moulded, PVC-free foam plugs are available in two sizes to ensure a proper and comfortable fit.
  • Polyurethane foam, without cord
  • Conical shape for easy insertion
  • Smooth outer surface for better hygiene
  • Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
Certifications: EN 352-2.
Packaging: Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.