Safety spectacles, uvex sportstyle
Supplier: UVEX
9193.376 9193.280 9193.838 9193.064 9193.265 9193.080
111-1704EA 29.9 EUR
111-1704 UVEX9193.280 UVEX9193.838 111-2388 111-2420 UVEX9193.080
Safety spectacles, uvex sportstyle
Safety Glasses
These light and comfortable, sports-style safety spectacles feature a wide field of vision with a contoured lens. They offer a good level of eye coverage and fit all facial shapes including narrower faces, which enables a standardisation of the range.
  • Due to significant anti-fog features these spectacles are suitable for high humidity environments
  • Permanent uvex lens coating technology (uvex supravision extreme): High performance anti-fog on the inside, scratch resistant on the outside
  • 100% UV400 protection
  • Adjustable nose bridge for comfortable