Disposable half masks against gas, vapours and particulates, FFA1/FFA2
Supplier: Moldex
5120 5230 5430 5330
111-1707EA 37.8 EUR
111-1707 111-1708 111-1710 111-1709
Disposable half masks against gas, vapours and particulates, FFA1/FFA2
Respirators Disposable Respirators Gas or Gas and Particulate Respirators
Maintenance-free compact half mask to protect against gas, vapour and dust.
  • Pre-assembled and ready for use
  • Lightweight dual-material construction
  • Adjusts to different face shapes automatically
  • Low profile design offers improved field of vision
Certifications: EN 405:2001