Earmuffs, uvex K2H
Supplier: UVEX
2600.202 2600.204
111-1905EA 38.5 EUR
111-1905 111-1924
Earmuffs, uvex K2H
Earmuffs Noise Blocking Earmuffs
These earmuffs for helmet mounting are suitable for use in medium noise areas (33 dB). They can be attached to the following uvex safety helmets: uvex pheos B-WR, uvex pheos alpine, uvex airwing and uvex superboss (additional adapter systems for combination with safety helmets and visors available on request). Two models are specifically designed for use with the uvex pheos helmet system.
  • Soft ear cushions adapt to surface and head shape to ensure optimal comfort
  • Length adjustment: Easily adjustable for a perfect fit on the helmet
  • 360° rotation for standby