Resusable particle filtering half masks, 8825+ and 8835+
Supplier: 3M
8825PLUS 8835PLUS
111-2293EA 66.4 EUR
111-2293 111-2326
Resusable particle filtering half masks, 8825+ and 8835+
Respirators Reusable Respirators Half Face Respirators
The 3M™ Particulate Respirator 8825+ has been engineered to help improve fit to wearer’s individual face shape and size. With it's robust shell and cup design, this respirator meets the particular requirements of the clogging test for FFP (filtering face piece) respirators.
  • Flexible material helps to improve fit to wearer’s individual face shape and size
  • High performance filter material that’s easy to breathe through
  • 3M™ Cool Flow™ Valve provides effective exit of heat build up provides a cooler and more comfortable wear and helps to minimise the risk of