Safety spectacles, uvex futura
Supplier: UVEX
9180.125
111-8017EA 35.6 EUR
111-8017
Safety spectacles, uvex futura
Safety Glasses
Classic model with excellent field of vision and an outstanding fit available in a range of sizes.
  • Four-position arm adjustment with integrated locking
  • Adjustable lens angle
  • Padded arms for greater comfort and a better fit
  • Article no. 111-8019 is ideal for slim faces
Weight: 46 g
Certifications: EN 166, EN 168, EN 170