Thulium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11199.04 11199.09
11199.04EA 221 EUR
11199.04 11199.09
Thulium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Thulium (III) oxide

Purity: 99.99% CAS No.: 12036-44-1 Molecular Formula: O3Tm2 Formula Weight: 385.87 Physical Form: Powder Appearance: Pale green Melting Point: 2425° Merck Reference: 14,9394 MDL No.: MFCD00011285

Formula: O₃Tm₂
Melting Pt: 2425 °C
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6
Merck Index: 14,9394

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR