Cleanroom gloves, polychloroprene, sterile, BioClean P-Zero™
Supplier: Nitritex
BPZS80 BPZS90 BPZS60 BPZS65 BPZS70 BPZS75 BPZS85
112-3877EA 622 EUR
112-3877 112-3879 112-3873 112-3874 112-3875 112-3876 112-3878
Cleanroom gloves, polychloroprene, sterile, BioClean P-Zero™
Gloves
These hand-specific gloves with beaded cuffs are resistant against chemicals and cytotoxic drugs. They are therefore suitable for chemotherapy applications (ASTM D 6978-05 tested).
  • Sterile, powder-free, textured finish for improved grip
  • Latex-free to eliminate Type I latex allergies
  • Beaded cuff provides easier donning and stability on the arm
  • Polychloroprene provides exceptional tactility
  • Excellent anti-static properties
Suitable for use in Class 10, ISO 4 or M2.5 and EU GMP Grade A cleanrooms.
EN 374-1/2/3, EN 455; Cat. III, AQL 1,5