Chemical resistant gloves, uvex profastrong NF33
Supplier: UVEX
6012201 6012202 6012204 6012203
112-4560EA 6.5 EUR
112-4560 112-4561 112-4563 112-4562
Chemical resistant gloves, uvex profastrong NF33
Gloves
These nitrile rubber gloves with cotton lining provide a high resistance against oils, greases, acids and solvents.
  • Abrasion resistant
  • Good grip, even on wet parts
  • Anatomical fit
  • Good dexterity
Ideal for use in the chemical, automotive, printing and food industry. Also suitable for laboratory applications.
Certifications: EN 388:2016 (4101X), EN ISO 374-1:2016 Type A (AJKLOT)
Food contact approved