Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK
97982 97984 97985
112-4867EA 0 EUR
112-4867 112-4869 112-4870
Nitrile gloves, Kimtech™ Blue Nitrile™
Gloves
These gloves are manufactured from nitrile instead of rubber latex, which reduces the likelihood of TYPE I glove-associated reactions.
  • Constructed of synthetic nitrile polymer and therefore free of silicone and latex
  • Smooth bisque finish enables excellent tactile sensitivity in the handling of both wet and dry materials and equipment
  • Beaded cuffs for added strength and easier donning and doffing
  • Compliant protection against viruses, chemical splash and microbiological hazards
  • Virus protection according to ISO 16604 procedure B:2004
  • Static dissipative