Safety helmet, uvex pheos planet B-S-WR
Supplier: UVEX
9772.043
112-6185EA 28.6 EUR
112-6185
Safety helmet, uvex pheos planet B-S-WR
Head Protection Safety Helmets
Lightweight helmets made from HDPE.
  • Suspension harness with wheel ratchet system for variable width adjustment
  • Safety helmet with helmet torch slot on the front
  • Euroslot adapters on the side (30 mm) for attaching earmuffs and the uvex pheos visor system
  • The shortened brim ensures unrestricted upwards vision
  • Six-point textile band attachment ensures excellent fit and comfort
  • Three adjustable air vents for maximum ventilation
Ideal for industry, road construction works, sewage and pipeline construction, concrete/construction work, brickwork,More Product Information