Cleanroom face masks, BioClean DB™, PE
Supplier: Nitritex
BDBS-G BDBS
113-0783EA 663 EUR
113-0783 113-0782
Cleanroom face masks, BioClean DB™, PE
Masks Cleanroom Masks
The sterile pouch style mask has knitted headbands and all seams are ultrasonically bonded. The mask has inner and outer facing layers constructed of spunbond non woven polyester with the middle filter layer made from meltblown polyester.
  • Spacer maintains a clear breathing space for the wearer, improving the overall comfort and ease of breathing
  • Plastic coated, soft malleable steel nose bridge is fully enclosed
  • Available with or without integral neck guard to close the gap in protection between the coverall and throat of the wearer