Chemical resistant overalls, KLEENGUARD* A71
Supplier: KIMBERLY CLARK
96760 96770 96790 96800 96780
113-6390EA 350 EUR
113-6390 113-6391 113-6393 113-6394 113-6392
Chemical resistant overalls, KLEENGUARD* A71
Disposable Garments
These disposable overalls made from a laminate of barrier film and a non woven fabric provide protection against high pressure aqueous jets, inorganic acids and bases and a range of organic chemicals. The non woven fabric is anti-static and silicone-free which is important when working with paint or sensitive surfaces.
  • Low lint and silicone-free for critical areas
  • Stitched and over-taped seams to provide high strength and barrier properties to withstand high pressure aqueous jets
  • With hood and zip closure with flap
Colour: Yellow
Certifications: More Product Information
Order Now
SPECIFICATIONS
Model A71
Environmentally Preferable
Closure type Zip closure
Anti-static +
Gender
Garment application Chemical protection,Paint and lacquer,Laboratory,Waste water,Industrial