Laboratory coats, PP
Supplier: HUBEI ORIENT INTL TRADING
113-8132EA 111 EUR
113-8132 113-8125 113-8126 113-8127 113-8128
Laboratory coats, PP
Disposable Garments
These disposable laboratory coats made from PP are ideal for non critical applications.
  • Elasticated wrist
  • Press stud fastening (four snaps)
  • Two pockets below the waist
Weight: 30 g/m²
Colour: White
Order Now
SPECIFICATIONS
Environmentally Preferable
Closure type Press studs / snaps
Pockets With pockets
Gender Unisex
Garment application Laboratory
Weight 30 g/m²