Cleanroom face masks, Kimtech™ M3
Supplier: KIMBERLY CLARK
62451 62452
113-8143EA 249 EUR
113-8143 113-8144
Cleanroom face masks, Kimtech™ M3
Masks Cleanroom Masks
Pleated masks that provide proven particle filtration to avoid contamination.
  • Thermalbond inner-facing provides high levels of comfort and good breathability
  • Latex- and silicone-free for reduced risk of toxic reactions such as skin irritation
  • Enclosed nose bridge to provide high comfort and minimised exposure
  • Outer breathable layer is made of low lint polyethylene
  • Particle filtration efficiency (PFE) > 96,9%
Packaging: Double bagged for cleanroom use.