VWR®, Cleanroom Face Masks, Duck Bill
Supplier: VWR Collection
113-8681EA 234 EUR
113-8681 113-8682 113-8683 113-8684 113-8685 113-8686
VWR®, Cleanroom Face Masks, Duck Bill
Masks Cleanroom Masks
Cleanroom, duck bill face masks feature superior particle and bacterial filtration efficiency plus low particle shedding. These moisture resistant, cleanroom masks consist of three layers featuring a unique, lightweight inner layer of film for enhanced moisture resistance.
  • Available as sterile or non sterile, with ear loops, ties or hood loops
  • High bacterial and particle filtration efficiency
  • Full length plastic arch, within the layers, holds the fabric away from the mouth to allow cooler, more comfortable breathing, without restricting downward vision