Safety boots, Wellingtons, Nora
Supplier: UVEX
9475.6/37 9475.6/39 9475.6/38 9475.6/40 9475.6/41 9475.6/42 9475.6/43 9475.6/44 9475.6/45 9475.6/46 9475.6/47 9475.6/48 9476.6/39 9476.6/37 9476.6/38 9476.6/40 9476.6/41 9476.6/42 9476.6/43 9476.6/44 9476.6/45 9476.6/46 9476.6/47
114-5830EA 39.1 EUR
114-5830 114-5832 114-5831 114-5833 114-5834 114-5835 114-5836 114-5837 114-5838 114-5839 114-5840 114-5841 114-8290 114-8288 114-8289 114-8291 114-8292 114-8293 114-8294 114-8295 114-8296 114-8297 114-8298
Safety boots, Wellingtons, Nora
Shoes and Boots
High PVC safety boots are anti-static, resistant to oil, grease, fuel, acids, alkalis and disinfectants.
  • Anatomical shape prevents foot fatigue
  • Long shaft can be shortend to meet individual requirements
  • Toe and heel reinforcement
  • Ankle protection and ribbed heel with kick-off tab to assist removal
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: yellow or white.
Shaft height: 330 mm.
Certifications: EN 20345:2011; S5, SRA