Cleanroom boot covers, CleanMax®
Supplier: LAKELAND CLEANMAX APPAREL
CTL903CMP-2X CTL903CMP-L/X CTL903CMP-S/M
114-9545EA 446 EUR
114-9545 114-9546 114-9547
Cleanroom boot covers, CleanMax®
Disposable Garments
Clean manufactured disposable boot covers are individually packaged in pairs and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • 19" high
  • Non-skid PVC sole
  • Individually bagged in pairs
  • Latex-free
  • Tunnelled elastic top
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparel provides chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borneMore Product Information