VWR® PureStep, Multi-Layer Adhesive Floor Mats
Supplier: VWR Collection
115-0047EA 104 EUR
115-0047 115-0048 115-0049 115-0050 115-0051 115-0052 115-0053 115-0054 115-0055 115-0056 115-0057 115-0058 115-0059 115-0060 115-0061 115-0062 115-0063 115-0064 115-0065 115-0066 115-0067 115-0068 115-0069 115-0070 115-0071 115-0072 115-0075 115-0076 115-0077 115-0078 115-0079 115-0080 115-0081 115-0082 115-0083 115-0084 115-0085 115-0086 115-0087 115-0088 115-0089 115-0090 115-0091 115-0092 115-0093 115-0073 115-0074 115-0167 115-0168 115-0169 115-1748 115-1774 115-1775 115-1776
VWR® PureStep, Multi-Layer Adhesive Floor Mats
Floor Mats Adhesive Floor Mats
30 or 60 layer adhesive entrance mats manufactured from low density polyethylene. These mats control contamination by removing debris from the soles of shoes, and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.
  • Antimicrobial agent protects against the growth of bacteria and fungi
  • Non slip, low profile, individually numbered layers
  • Reduced maintenance and cleaning costs
  • Contains GMO-free, renewable plant-based material that reduces CO₂ emissions
  • Adhesive mats are