Cleanroom wipes, BetaWipe™
Supplier: Texwipe
TX2009
115-0304EA 608 EUR
115-0304
Cleanroom wipes, BetaWipe™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
This thick polypropylene and cellulose wipe is thermally bonded in a unique composite construction. The cellulose contributes to the product’s high sorption, while the PP provides the wipe with resistance to strong acids. With its very soft surface texture, it's is ideal for cleaning sensitive surfaces and delicate instruments.
  • Provides an extremely soft and no nabrasive surface texture
  • Low particle generation helps prevent contamination
  • Designed for removal of acids, etchants and chemical spills
  • Autoclavable
Suitable for use in Class 1000, ISO 6, M4.5More Product Information