Cleanroom wipes, Vectra® AlphaSorb® 10 and AlphaSorb® 10
Supplier: Texwipe
TX1052 TX1050
115-0603EA 1340 EUR
115-0603 115-1788
Cleanroom wipes, Vectra® AlphaSorb® 10 and AlphaSorb® 10
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These 2-ply double-knit 100% continuous-filament polyester wipes offer high sorbency performance.
  • All four edges are thermally sealed, forming a border to prevent particle and fibre release
  • Assured cleanliness with ultralow levels of particles and extractables
  • Unique 2-ply, pinsoniced construction for extended durability
  • Vectra® AlphaSorb® 10 wipes are Vectra processed and AlphaSorb® 10 wipes are laundered in a cleanroom
Ideal for use in the most critical cleanroom environments, such as semiconductor and pharmaceutical manufacturing. They are alsoMore Product Information