VWR® Absorbent Bench Underpads
Supplier: VWR Collection
56616-030 56616-031 56616-018
115-0683EA 234 EUR
115-0683 115-0684 VWRU56616-018
VWR® Absorbent Bench Underpads
Surface Protectors Bench Protectors
Absorbent underpads with a three-layer construction made from PP top and back sheets and a cellulose filler.
  • Top sheet made from white spunbound polypropylene
  • Absorbent cellulose fluff filler between two layers of tissue for superior wicking of fluid
  • Cushion effect to protect labware from breakage
  • Embossed PP backing sheet comes in different colours
  • Sealed on four sides to prevent liquid seepage
Deluxe pads are more absorbent and thicker with more cellulose fluff than standard pads.
Ordering information: Super Deluxe pads have an added polymer that absorbs more moisture.