Cleanroom anti-fatigue mats, Ergomat® Nitril ESD Conductive
Supplier: ERGOMAT
ENC0306 ENC0420 ENC0412 ENC0320 ENC0304 ENC0220 ENC0209 ENC0206 ENC0406 ENC0312 ENC0212 ENC0303 ENC0309 ENC0204 ENC0404 ENC0409
115-0880EA 600 EUR
115-0880 115-0884 115-0883 115-0881 115-0879 115-0878 115-0877 115-0876 115-0882 ERGOENC0312 ERGOENC0212 ERGOENC0303 ERGOENC0309 ERGOENC0204 ERGOENC0404 ERGOENC0409
Cleanroom anti-fatigue mats, Ergomat® Nitril ESD Conductive
Floor Mats Traction and Anti-Fatigue Floor Mats
The Ergomat® Nitril ESD Conductive is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave. It is ideal for usage in the electronics industry or for areas where static control is crucial due to sensitive equipment.
  • Upper side with cones
  • Great anti-bacterial cleaning properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantee durability