Protective ointment, Travabon® classic
Supplier: Deb-STOKO
22325 TVC100ML TVC1L
115-1471EA 345.69 EUR
115-1471 115-2379 115-2089
Protective ointment, Travabon® classic
Skincare Skin Creams Hand Creams
Specialist pre-work skin protection cream against stubborn, non water-soluble dirt such as oils, paints, graphite, metallic dust, adhesives and water-insoluble workplace substances. Suitable for use on hands and body. For areas where gloves are not worn.
  • Facilitates cleansing of the skin thanks to soil suspending emulsifiers
  • Enhanced protective action through skin protection pigments
  • With glycerine and emollients
  • Water-soluble, oil-free, colourant-free, silicone-free, perfumed
Ordering information: Soft bottle may only be used with Stoko® Vario®More Product Information