Multi-layer adhesive floor mats for cleanrooms, CleanStep™
Supplier: Texwipe
AMA184681B AMA184681W AMA184682G AMA184682W AMA254581W AMA254582W AMA364681W AMA254581B AMA364681B AMA184682B
115-0201EA 211 EUR
115-0201 115-0136 115-2695 115-0135 115-0132 115-0131 115-0137 115-2696 115-2698 115-2694
Multi-layer adhesive floor mats for cleanrooms, CleanStep™
Floor Mats Adhesive Floor Mats
Adhesive coated contamination control mats capture particulates from foot traffic and equipment wheels, providing the most effective dirt and dust removal method available in a mat.
  • Self-adhesive base prevents slipping
  • Consistent adhesive coating on each layer
  • Anti-microbial; reduces the potential of spreading germs and bacteria
  • Low profile (approx. 1,5 mm for 30 layers, 2 mm for 60 layers) prevents tripping
  • Each layer is numbered, so the number of layers remaining is easily identified
  • Clear film layers over colour bottom film for colour consistency