Dual-wire thermoplastic wrist strap
Supplier: DESCO
4720
115-3067EA 10.1 EUR
115-3067
Dual-wire thermoplastic wrist strap
Grounders Wrist Grounders
The Dual-Wire thermoplastic wrist strap consists of an insulative exterior with a conductive interior and a 3,4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
  • Zipper style latching mechanism easily adjusts to most wrist sizes
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of EN 61340-5-1 and Wrist Strap standard IEC 61340-4-6.More Product Information