Palladium ≥99.9% (metals basis), foil, Thickness: 1.0 mm

Supplier: Thermo Fisher Scientific

11513.FF 11513.FI
11513.FFEA 2520 EUR
11513.FF 11513.FI
Palladium ≥99.9% (metals basis), foil, Thickness: 1.0 mm
Palladium

Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈7.5g/25x25mm Odour: Odourless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR