SPINeasy® DNA Kit for Microbiome
Supplier: MP Biomedicals
116553050
116553050.EA 233 EUR
116553050.
SPINeasy® DNA Kit for Microbiome
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
SPINeasy® DNA Kit for Microbiome employs a novel method for isolating microbial genomic DNA from various samples. Due to its highly effective lysis capability and silica-membrane spin-column technology, the SPINeasy® DNA Kit for Microbiome can be used for bacterial, body fluids and environmental samples such as soil and stool.
  • For genomic, molecular biology, PCR, qPCR, microbiome analysis, NGS, Shotgun sequencing
  • Personal protective equipment: Safety goggles/face shield, gloves
With the use of specially formulated Buffer MB1 and Lysing Matrix E in combination with FastPrep® Instruments from MP Biomedicals, an efficient lysis of various samples can be achieved within seconds. Provided in the kit, the Column MB and kit buffers are designed to deliver gDNA of high yield and purity, and compatible with downstream applications such as qPCR, restriction digestion, and sequencing.
Efficient and unbiased lysis of microbes, including gram positive/negative bacteria, fungi, protozoans, and viruses found in swab, sputum, body fluid, stool, soil, and water samples.
SPINeasy® DNA Kit for Microbiome employs a novel method for isolating microbial genomic DNA from various samples. Due to its highly effective lysis capability and silica membrane spin column technology, the SPINeasy DNA Kit for Microbiome can be used for bacterial, body fluids, and environmental samples such as soil and stool.